CPS Products LS790B Professional Refrigerant Leak Seeker
Representative image only.
CPS Products Mfr # LS790B – Item # 1011218
Unique "Gearshift" LED display for leak sizing allows user to determine the approximate size of a leak
  • 2 year warranty
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Weight (lbs)1.016
  • Weight (grams)500
  • Battery TypeThree C alkaline batteries
  • Battery Life75 hours
  • DescriptionProfessional Refrigerant Leak Seeker
  • Warranty2 year
Two year warrantyConformite Europeenne
Key Features
  • 2 year warranty
More About this Item
The advanced line of CPS® leak detectors combine the most advanced microcomputer circuitry with unique sensor technology to create highly sensitive, accurate electronic handheld leak detection devices. All units have multiple visual and audible alarm indicators to quickly alert the user to varying halogen concentrations. Unique automatic calibration design eliminates the need for manual balance or tick rate adjustment.
LS790B includes automatic calibration which records background levels of refrigerant, eliminating the need for manual adjustments. Features over 100 computer generated audible tones and a bar graph display for leak indication.