Diba Omnifit® Activated Carbon Vapor Trap for Solvent Waste Caps with Venting Port; 5/pk
Representative image only.
Diba Mfr # 009CF-TWB – Item # 2194331
Prevent chemical vapors from escaping from solvent containers
  • Compatible with Diba T-series solvent caps
  • Connects to 1/4"-28 UNF vent port
PRODUCT OPTIONS
₹232,000.00INR / Pkg of 5
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithT-series solvent caps with vent; 1/4"-28 UNF(M) connection
Key Features
  • Compatible with Diba T-series solvent caps
  • Connects to 1/4"-28 UNF vent port
More About this Item
Use traps with Luer connection with T-series solvent waste caps without valve and filter. Traps with 1/4"-28 UNF(M) connection works with caps with vent. Traps with UNF connection feature an indicator strip that changes color as it gets saturated.
Use activated carbon trap with T-series solvent caps with vent port. An indicator strip changes color to show when the vapor trap is saturated.