Diba Omnifit® EZ Column Replacement Glass, 6.6 mm ID x 100 mm L
Representative image only.
Diba Mfr # 006ERG-06-10 – Item # 1194228
Mix and match column length while using your existing endpieces for ultimate flexibility
  • Constructed of durable glass
PRODUCT OPTIONS
₹21,300.00INR / Each
Include Column Today
SPECS & DESCRIPTION
Specifications & Description
  • Column Length (mm)100
  • Column Inside Diameter (ID) (mm)6.6
  • Wetted MaterialsGlass
  • Compatable WithOmnifit® EZ and EZ SolventPlus™ Column 6.6 mm ID x 100 mm L
Key Features
  • Constructed of durable glass
More About this Item
Use a different length tube with your existing endpieces to meet your specific requirements. Columns sold separately.