Diba Omnifit® EZ Packing Sleeve, Join 2 Columns or Connect to Column Packer, 10 mm; 1/ea
Representative image only.
Diba Mfr # 006EPS-10 – Item # 1194216
Use packing sleeve to join two columns for filling and packing
  • Suitable for use with Diba Omnifit® EZ and EZ SolventPlus™ columns
PRODUCT OPTIONS
₹10,800.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Column Inside Diameter (ID) (mm)10
Key Features
  • Suitable for use with Diba Omnifit® EZ and EZ SolventPlus™ columns
More About this Item
These Diba Omnifit® packing sleeves allow you to easily join two columns or connect a column to a column packer. Choose from a wide range of column inner diameter sizes suitable for your application.