Digi-Sense Data Logging Vane Anemometer with NIST-Traceable Calibration

Representative image only.
Digi-Sense Item # 2025022
Versatile handheld or mountable design for quick air velocity sampling or long term monitoring
  • Reliable measurements, guaranteed! Precalibrated to NIST-traceable standards.
  • User-selectable units of measure eliminate the need for conversions
  • Measure air velocities up to 3937 ft/min (20.0 m/s) to cover broad range of applications
  • Analyze and graph logged data using included software
  • User-defined high and low alarm levels with visual LED indication to bring attention to potential problems
  • Articulated screw-on mounting stand provides flexible positioning to fit the application
  • 2 year warranty

Call For Price
Time to recalibrate?
for an additional
₹45,100.00INR / Each
for an additional
₹55,300.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Max Air Velocity (ft/min)3837
  • Min Air Velocity (ft/min)80
  • Air Velocity Resolution1 ft/min (0.01 m/s, 0.1 km/hr, 0.1 mph, 0.1 knots)
  • Air Velocity Accuracy±(3% + 40 ft/min or 0.2 m/s)
  • Data LoggingYes
  • Data Storage (samples)32000
  • Logging Intervals2, 5, 10, 30 seconds, 1, 5, 10, 30 minutes, 1, 2, 3, 6, 12, 24 hours
  • Width (in)1 3/8
  • Height (in)1 1/8
  • Length (in)5 3/4
  • Battery1 3.6V lithium (included)
  • Warranty2 year
Two year warrantyConformite EuropeenneNIST Traceable Calibration

Key Features

  • Reliable measurements, guaranteed! Precalibrated to NIST-traceable standards.
  • User-selectable units of measure eliminate the need for conversions
  • Measure air velocities up to 3937 ft/min (20.0 m/s) to cover broad range of applications
  • Analyze and graph logged data using included software
  • User-defined high and low alarm levels with visual LED indication to bring attention to potential problems
  • Articulated screw-on mounting stand provides flexible positioning to fit the application
  • 2 year warranty

More About this Item

The Digi-Sense data logging anemometer is designed for precision air velocity measurements in five user-selectable units (ft/min, m/s, mph, km/h, or knots). The instrument will help you meet cooling and environmental requirements for safety and comfort. A metallic mounting stand is included that provides an opportunity to position the anemometer to best fit the application. Built-in USB interface and included software allow logged data to be easily downloaded to a PC for analysis and graphing.
Digi-Sense comes pre-calibrated, saving you hundreds of dollars and extra waiting time to calibrate separately

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat