Digi-Sense PFA Safety Coated Liquid-In-Glass Thermometer; 0 to 230F, Total Immersion, Organic Liquid Fill
Representative image only.
Digi-Sense Item # 0807783
The liquid and glass are contained within the PTFE jacket, preventing contamination
  • Nonmercury blue spirit liquid
  • Statement of accuracy
  • 5 year warranty
Stay connected — Monitor your critical environments
SPECS & DESCRIPTION
Specifications & Description
  • Length (")12
  • Min Temperature (° F)0
  • Max Temperature (° F)230
  • Accuracy±3.0°F; ±4.0°F above 212°F
  • Division (° F)2
  • Thermometer FillBlue Spirit
  • Overall Length (InchShort)12
  • Overall Length (mm)300
  • Immersion Depth (mm)300
  • Stem MaterialPFA-Coated White-Back Glass
  • Warranty5 years
5 Year Warranty
Key Features
  • Nonmercury blue spirit liquid
  • Statement of accuracy
  • 5 year warranty
More About this Item
These thermometers are ideal for use in laboratories, universities, food/beverage, environmental, wastewater, and many other general-purpose applications. In the event of breakage, the PTFE coating contains the broken liquid and glass, preventing contamination or injury.
Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered.