Digi-Sense Pocket Liquid-In-Glass Thermometer; -30 to 120F, Closed Metal Case, Organic Liquid Fill
Representative image only.
Digi-Sense Item # 9025016
Low-cost way to measure temperature with a compact and durable metal case
  • Non-mercury blue spirit liquid
  • Closed metal case
  • 1 year warranty
PRODUCT OPTIONS
₹4,800.00INR / Each
₹34,000.00 - ₹43,100.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
Stay connected — Monitor your critical environments
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)-30
  • Length (")6 19/64
  • Max Temperature (° F)120
  • Accuracy±2.0°F
  • Division (° F)2
  • Thermometer FillBlue Spirit
  • Dial / Case MaterialClosed Metal
  • Overall Length (InchShort)6.3
  • Overall Length (mm)160
  • Stem MaterialWhite-Back Glass
  • Warranty1 year
1 Year Warranty
Key Features
  • Non-mercury blue spirit liquid
  • Closed metal case
  • 1 year warranty
More About this Item
These thermometers are ideal for use in laboratory, biology, chemistry, university, photographic, food/beverage, petrochemical, automotive, wastewater and many other general purpose applications. The protective plastic case protects these thermometers from accidental breakage during use and storage. The section of the case that surrounds the bulb has small openings for circulation. All DURAC Plus® thermometers feature markings that are permanently printed on the glass, blue spirit liquid that has a dark scale contrasts against a white background.