Digi-Sense Solar Twin Temperature Industrial RTD Probe; 9" Stem, -50-300F
Representative image only.
Digi-Sense Item # 9382059
An innovative, cost effective way to solve two types of temperature measuring needs while still only using one entry point into your process.
  • 1 year warranty
₹112,000.00INR / Each
₹17,000.00 - ₹25,900.00INR / Each
for an additional
₹17,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeRTD
  • Min Temperature (° F)-50
  • Max Temperature (° F)300
  • Min Temperature (° C)-46
  • Max Temperature (° C)149
  • Connector Type1/2" NPT(M)
  • Sheath Length (in)6
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Cole Parmer solar Twin Temp industrial thermometers are the perfect solution for systems requiring local and remote temperature readings. Local readings are provided by digital screen. The remote reading is achieved with a continuou 4-20 mA output signal. All sensors are located in the thermometer stem and will easily fit into existing bi-metal thermowells of corresponding length. The housing is manufactured of 304 stainless and is guaranteed to be hermetically sealed to prevent fogging.