Digi-Sense TC/PR28875-3 TC/PR28875-3 Type K 90°-Angle Surface Thermocouple Probe with Guarded Tip

Representative image only.
Digi-Sense Item # 0844510
Surface Probes with PTFE guard are designed to measure surface temperature without penetrating the surface
  • These probes are designed to measure surface temperature without penetrating the surface
  • Ideal for monitoring flat surfaces such as hot plates, furnaces, or molds
  • Each probe comes with a PTFE guard to protect the junction and help keep the temperature reading isolated
  • Response Time: 1 second or greater
  • Each probe comes with a color coded ANSI coiled cable: type J-black, type K-yellow, type T-blue, and type E-purple.
  • Custom designs or changes to this product are available upon request
  • 4 month warranty

₹26,200.00INR / Each

₹34,000.00 - ₹43,100.00INR / Each

for an additional
₹34,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
Often Bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Length (")5
  • Probe TypeK
  • Min Temperature (° F)-418
  • Max Temperature (° F)500
  • Min Temperature (° C)-250
  • Max Temperature (° C)260
  • Time Constant1 second
  • JunctionIsolated
  • Connector TypeMini Connector
  • Sheath Length (in)5
  • Sheath Diameter (in)0.5
  • Warranty4 months

Key Features

  • These probes are designed to measure surface temperature without penetrating the surface
  • Ideal for monitoring flat surfaces such as hot plates, furnaces, or molds
  • Each probe comes with a PTFE guard to protect the junction and help keep the temperature reading isolated
  • Response Time: 1 second or greater
  • Each probe comes with a color coded ANSI coiled cable: type J-black, type K-yellow, type T-blue, and type E-purple.
  • Custom designs or changes to this product are available upon request
  • 4 month warranty

More About this Item

Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat