Digi-Sense Temp-300 Dual-Input Data Logging Thermocouple Thermometer
Representative image only.
Digi-Sense Item # 9142804
Heavy-duty thermometer offers versatility and accuracy in a compact design
  • 3-way hands-free operation available
  • Large easy-to-read screen with back light
  • Rugged armor with built-in stand—withstand the toughest conditions
  • Easy-to-use automatic field calibration ensures accurate readings
  • Probes are sold separately
  • 3 year warranty
PRODUCT OPTIONS
₹122,000.00INR / Each
₹23,000.00 - ₹32,800.00INR / Each
for an additional
₹32,800.00INR / Each
for an additional
₹23,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeThermocouple
  • Number Of Probe Channels2
  • Types Of Probes AcceptedType B, Type E, Type J, Type K, Type N, Type R, Type S, Type T
  • Min Temperature (° F)-418
  • Max Temperature (° F)3214
  • Min Temperature (° C)-250
  • Max Temperature (° C)1768
  • AccuracyType E, J, K, N, T: ±0.1% of reading +0.7°F (+0.4°C); ±0.25% of reading +2.0°F (+1.0°C) below -148°F (-99.9°C); Type R, S, B: ±0.1% of reading +2.0°F (+1.0°C)
  • Resolution0.1°; 1° above 999.9° and below -99.9°
  • OutputUSB
  • Data LoggingYes
  • Memory Capacity2000 data points
  • IP RatingIP54
  • Low Battery IndicatorYes
  • Display Type1.5" Display, 4-digit LCD
  • Height (in)7
  • Depth (in)4
  • Battery Life400 hours
  • Battery3 AA (included)
  • DescriptionTemp-300 Dual-Input Datalogging Thermocouple Thermometer with Boot
  • Warranty3 year
3 Year WarrantyConformite Europeenne
Key Features
  • 3-way hands-free operation available
  • Large easy-to-read screen with back light
  • Rugged armor with built-in stand—withstand the toughest conditions
  • Easy-to-use automatic field calibration ensures accurate readings
  • Probes are sold separately
  • 3 year warranty
More About this Item
These robust and reliable meters work in any environment from the lab to the production floor. The sturdy armored construction protects against knocks and jars. All models are ergonomically designed for both large and small hands alike. The sealed keypad and ABS plastic case meet IP54 standards for splash resistance. Select from the three most popular thermocouple types to meet all your basic temperature measurement needs.
Digi-Sense's exclusive “3-WAY HANDS-FREE” capability allows “hands-free” use—use the built-in stand on a table top, attach the meter to metal objects through magnets inserted into the armor boot, or hang from a pipe or belt using the hook-and-loop strap. Meters come standard with the integral stand; the “hands-free” option adds magnets and strap (order separately).
The Digi-Sense® Temp-300 thermometer features real-time data logging and a USB data output. Meter accepts dual inputs from type J, K, T, E, R, S, N, or B thermocouples probes with ANSI miniconnector. This robust and reliable meter works in any environment from the lab to the production floor. Sturdy armored construction protects against knocks and jars. Sealed keypad and ABS plastic case meet IP54 standards for splash resistance. Ergonomically designed housing is ideal for both large and small hands alike.
Large, illuminated display features easy-to-read digits. The seven-button control panel and intuitive on-screen menus guide you in choosing your desired functionality. Additional meter features include Min/Max, Hold, and average functions; temperature units in °F, °C, °R, or K; offset; time stamp/clock; countdown timer; out-of-range alarm with audible/visible indicator; auto shutoff; and low-battery indicator. Order an optional AC adapter to conserve battery life during extended operations.