Digi-Sense Thermocouple Wire Type K 20-Gauge, High Temperature Glass Braid, 100ft

Representative image only.
Digi-Sense Item # 0854123
Make your own extension cables
  • 6 month warranty

PRODUCT OPTIONS

₹25,200.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • MaterialHigh Temp Fiberglass
  • Wire Gauge (Ga)20
  • Error Limit±2% reading from -330 to -165°F (-200 to -110°C); ±4°F (±2.2°C) from -165 to 545°F (-110 to 285°C); ±0.75% reading from 545 to 2300°F (285 to 1250°C)
  • Inner InsulationHigh Temp Fiberglass Braid
  • Outer InsulationHigh Temp Fiberglass Braid
  • Stranded / SolidSolid
  • OverbraidNo
  • Special InformationSpecial limits of error (±4%)
  • Min Temperature (° F)0
  • Max Temperature (° F)1400
  • Min Temperature (° C)-18
  • Max Temperature (° C)750
  • Length (ft)100
  • Size (in)0.080 in x 0.136 in
  • Warranty6 month
6 Month Warranty

Key Features

  • 6 month warranty

More About this Item

Wire comes in 20-gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with polyvinyl chloride (PVC), fluorinated ethylene propylene resin (FEP/FEP), fiberglass, high-temperature fiberglass, or ceramic fiber outer/inner insulation. These cables are also available in 100-ft or 1000-ft lengths for any application.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat