Digi-Sense Type-J Self-Adhesive Tape Probe 1" x 2" Mini-Con 5ft FEP Ins Wire (Pk/3)

Representative image only.
Digi-Sense Item # 0851950
Self-Adhesive Probes Designed for Easy Installation and Removal on a Wide Range of Surfaces
  • The Kapton tape is rated to 400°F (203°C)
  • Tape can be applied and to almost any surface
  • Response Time: 1.5 seconds or greater
  • Each probe comes with a color coded ANSI mini connector: type J-black, type K-yellow, type T-blue, and type E-purple
  • Custom designs or changes to this product are available upon request
  • 6 month warranty

PRODUCT OPTIONS

₹12,500.00INR / Pkg of 1

₹34,000.00 - ₹43,100.00INR / Each

for an additional
₹34,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Probe TypeJ
  • Min Temperature (° F)32
  • Max Temperature (° F)675
  • Min Temperature (° C)0
  • Max Temperature (° C)357
  • Temperature Accuracy±5.9°F (±3.3°C), or ±0.4% of reading above 32°F (0°C); ±4.0% of reading below 32°F (0°C)
  • JunctionGrounded
  • Connector TypeMini Connector
  • Sheath materialKapton Tape glue
  • Cable Length (in)60
  • Warranty6 month
6 Month Warranty

Key Features

  • The Kapton tape is rated to 400°F (203°C)
  • Tape can be applied and to almost any surface
  • Response Time: 1.5 seconds or greater
  • Each probe comes with a color coded ANSI mini connector: type J-black, type K-yellow, type T-blue, and type E-purple
  • Custom designs or changes to this product are available upon request
  • 6 month warranty

More About this Item

Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).black, type K—yellow, type T—blue, and type E—purple. Probes with a plastic handle have a maximum temperature of 275°F (135°C)
before being affected.These self-adhesive probes adhere to most surfaces. Kapton-insulated wire and industrial adhesives for high temperature and long-term durability. Includes 5-ft. of straight thermocouple wire with miniconnectors.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat