Digi-Sense Type-K, Kapton-Insulated Probe, Mini-Connector, Exposed, 5ft 30 -Gauge, 6 Pk

Representative image only.
Digi-Sense Item # 0850586
Flexible Kapton-Insulated Probe for Faster Response Time than Most Insulated Wire Probes. Ideal for Checking Food Temperatures
  • Sold in packs of six.
  • Response Time: 0.35 seconds or greater
  • All insulated-wire probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors: type J - black, type K - yellow, and type T - blue.
  • Custom designs or changes to this product are available upon request
  • 6 month warranty

PRODUCT OPTIONS

₹16,800.00INR / Pkg of 6

₹34,000.00 - ₹43,100.00INR / Each

for an additional
₹34,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Length (")60
  • Probe TypeK
  • Min Temperature (° F)-250
  • Max Temperature (° F)600
  • Min Temperature (° C)-150
  • Max Temperature (° C)315
  • Temperature Accuracy±3.0°F (1.1°C) or ±0.4% reading above 212°F(100°C); ±3.6°F (2.2°C) or 2% of reading between -58 to 211°F (-50 to 99°C); ±7.2°F (4°C) or ±4% of reading between -60 to -238 (-51 to -150°C), whichever is greater
  • JunctionExposed
  • Connector TypeMini Connector
  • Sheath materialKapton Insulated Wire
  • Sheath Length (in)60
  • Sheath Diameter (in)0.01
  • Cable Length (in)60
  • Cable Wire Gauge (Ga)30
  • Warranty6 month
6 Month Warranty

Key Features

  • Sold in packs of six.
  • Response Time: 0.35 seconds or greater
  • All insulated-wire probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors: type J - black, type K - yellow, and type T - blue.
  • Custom designs or changes to this product are available upon request
  • 6 month warranty

More About this Item

Exposed-junction probes are not recommended for use with aqueous solutions.

Do not subject type J exposed-junction probes to moisture as they will rust.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat