DWK Life Sciences (Kimble) HPLC Mobile Phase Reservoir, Glass, Flat Bottom; 1.0 Liter
Representative image only.
DWK Life Sciences (Kimble) Mfr # 953902-1002 – Item # 0201720
Chemically inert glass prevents leaching of any extractables into the mobile phase solvents
  • Constructed of durable safety coated glass
  • 1 year warranty
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Bottom TypeFlat
  • Total Volume Capacity (mL)1000
  • Warranty1 year
1 Year Warranty
Key Features
  • Constructed of durable safety coated glass
  • 1 year warranty
More About this Item
These reservoirs are manufactured of high-quality borosilicate glass for maximum chemical resistance. Reservoirs are coated with plastic for safety and UV resistance. Select from conical bottom configuration for maximum solvent utilization or flat bottom reservoirs.