DWK Life Sciences (Wheaton) W224100-181 Vial Stopper, Snap-On, Gray Chlorobutyl, 20 mm; 1000/Cs

Representative image only.
DWK Life Sciences (Wheaton) Mfr # W224100-181 – Item # 0891902
Reliably seal vials
  • Choose from snap-on or straight plug vial stopper designs

PRODUCT OPTIONS

₹101,000.00INR / Case of 1000
Cole-Parmer Butyl Injection Stopper, 20 mm; 1000/pk
₹20,500.00INR / Pkg of 1000

SPECS & DESCRIPTION

Specifications & Description

  • Top Outside Diameter (OD) (mm)20
  • ColorGray
  • MaterialBromobutyl
  • DescriptionStopper, 20mm, Snap-On, Gray Butyl, 1000/CS

Key Features

  • Choose from snap-on or straight plug vial stopper designs

More About this Item

Straight plug style stoppers provide maximum sealing for vials with straight wall glass finish. The straight wall interior finish is typically found in vials from European manufacturers. This style is not recommended for vials with a blow-back feature, which is a small ridge on the interior of the vial's neck.

Snap-on style stoppers provide maximum sealing for vials with a a blow-back glass finish, which is a small ridge on the interior of the vial's neck. The snap-on stopper locks into the blow-back ridge, providing a tight seal and keeping the stopper from backing out. This style is not recommended for vials with a straight-walled finish. The blow-back feature is typically found in vials from North American manufacturers.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat