Edwards A133-05-000 Vacuum Pump Accessories, Inlet Foreline Trap, NW 25
Representative image only.
Edwards Mfr # A133-05-000 – Item # 7930220
  • 1 year warranty
₹144,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionVacuum Pump Accessories, Inlet Foreline Trap, NW 25
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Inlet foreline trap prevents back migration of rotary pump oil vapor into your inlet jet. Wetted parts are aluminum and VITON; 20 cfm max flow. Temp range is 32 to 122°>F. NW 25 ports. Measures 5-1/8"L x 3-1/4" dia. Includes activated alumina.