EMD Millipore UFC500324 Amicon Ultra Centrifugal Filters, 3 kDa Ultracel, 0.5 mL; 24/Pk
Representative image only.
EMD Millipore Mfr # UFC500324 – Item # 2995046
Highest recovery of sample with solute recoveries typically > 95%
  • Unique vertical housing
  • 1 year warranty
Loading
₹75,100.00INR / Each
Loading
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAmicon Ultra Centrifugal Filters, 3 kDa Ultracel, 0.5 mL; 24/Pk
  • Warranty1 year
1 Year Warranty
More About this Item
Ideal for dilute (ng/mL to µg/mL range) protein solutions. Low-binding ultracel-YM regenerated cellulose membrane available with different molecular weight cut-offs. Used in standard 1.5 mL tube fixed-angle rotors. Applications include concentrating and purifying proteins, antibodies and nucleic acids (alternative to EtOH precipitation), desalting and buffer exchange, and removal of primers, linkers and unin-corporated label.
Maximum initial sample volume is 500 µl, and typical final concentrate volume is 5 to 15 µl. Maximum centrifugal force is 14,000g , 500g for 29950-54 on DNA/RNA applications. Maximum initial sample volume is 500 ul, and typical final concentrate volume is 5 to 15 ul. Maximum centrifugal force is 14,000g , 500g for 29950-54 on DNA/RNA applications.