Environmental Express Environmental Express/ Oakton® pH electrode refillable double-junction for Orion A series meters
Representative image only.
Environmental Express Item # 2750830
Works with meters you already have
  • Combine the pH reference, pH half-cell, and temperature sensor in one 12-mm diameter electrode body
  • 1 year warranty
PRODUCT OPTIONS
₹31,800.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsRefillable
  • Electrode TypeDouble junction
  • Electrode Body MaterialEpoxy
  • ConnectorBNC
  • DescriptionpH electrode refillable double-junction for Orion A series meters
  • Warranty1 year
1 Year Warranty
Key Features
  • Combine the pH reference, pH half-cell, and temperature sensor in one 12-mm diameter electrode body
  • 1 year warranty
More About this Item
Epoxy-body pH electrodes compatible with Orion A-Series meters.