Environmental Express pH half-cell, general purpose, BNC
Representative image only.
Environmental Express Item # 0599015
Combine the half-cell and reference electrodes to create a system tailored to meet your specific application requirements
  • Refillable reference electrodes allow for repeated uses
  • 6 month warranty
₹22,900.00INR / Each
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsGeneral-purpose measurements
  • Electrode TypeHalf-cell; single junction
  • Electrode Body MaterialGlass
  • Max Temperature (° C)100
  • ConnectorBNC
  • Diameter (mm)12
  • Length (mm)154
  • Warranty6 month warranty
6 Month Warranty
Key Features
  • Refillable reference electrodes allow for repeated uses
  • 6 month warranty
More About this Item
These half-cell electrodes have BNC connectors and must be used with reference electrodes, available as 05990-60 or 05899-25. Both of these electrodes are made of blue sensing glass, have a 212°F (100°C) maximum temperature and are 154 mm long by 12 mm in diameter. Model 05990-15 is for general purpose pH use and 05990-16 is for general purpose ORP measurment.
Note: half-cell electrodes must be used with reference electrodes.