Eppendorf 952010051 Vis Disposable Cuvettes, PCR Clean, Protein Free; 80/Bx
Representative image only.
Eppendorf Mfr # 952010051 – Item # 8360051
Designed for easy and efficient molecular analyses
  • Optimize performance of your Eppendorf spectrophotometer with these accessories
  • Individually packaged and certified PCR clean and protein-free
  • UV transparent plastic, free of fluoropolymers and other halogenated hydrocarbons
  • Deep-lying optical window prevents scratches
  • Tapered cuvette base for optimal filling, frosted grip for labeling
PRODUCT OPTIONS
₹39,100.00INR / Box of 80
SPECS & DESCRIPTION
Specifications & Description
Key Features
  • Optimize performance of your Eppendorf spectrophotometer with these accessories
  • Individually packaged and certified PCR clean and protein-free
  • UV transparent plastic, free of fluoropolymers and other halogenated hydrocarbons
  • Deep-lying optical window prevents scratches
  • Tapered cuvette base for optimal filling, frosted grip for labeling
More About this Item
Improve your molecular anayses with these quality accessories.
These cuvettes feature two built-in optical path lengths in a single cuvette - just turn the UVette 90° to change from 10 mm and 2 mm.