Eppendorf Filter Tips, 50-1000 20 µL

Representative image only.
Eppendorf Mfr # 0030078578 – Item # 1888830
Protect your pipettor and prevent cross-contamination

PRODUCT OPTIONS

₹58,100.00INR / Case of 960

₹17,600.00 - ₹25,900.00INR / Each

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • SterileYes
  • Max Volume (µL)1000
  • Min Volume (µL)500
  • Min Volume (mL)0.5
  • Max Volume (mL)1
  • Tip Volume (mL)1
  • Min Dispensing (mL)0.5
  • Max Dispensing (mL)1
  • Color CodeBlue
  • Package Of10 racks; 96 tips per rack

More About this Item

Integrated hydrophobic filters do not contain "self-sealing" additives. All filter tips are sterile and PCR clean (free of DNA, DNase, RNase, and PCR inhibitors). Packaged in racks of 96 tips.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat