Excel Scientific PDL-5 Microplate Sealing Film Plate Paddle, Polyethylene, 3-7/8" x 2-15/16", White; 50/Cs
Representative image only.
Excel Scientific Mfr # PDL-5 – Item # 1302498
Easily adhere sealing films to microplates
  • Optimize performance of your microplate applications with these accessories
PRODUCT OPTIONS
₹16,400.00INR / Case of 50
Often Bought with…
Often Bought with…
Excel Scientific AlumaSeal Sealing Film
15,600.00 - ₹21,000.00INR / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Plate TypeSealing Film Paddle
  • Plate ColorWhite
  • MaterialPolyethylene
  • Width (in)3 7/8
  • Length (in)2 15/16
Key Features
  • Optimize performance of your microplate applications with these accessories
More About this Item
Ensure a secured seal of your sealing film with these accessories.
Complete the application of AlumaSeal 96 sealing films by pressing with the film-sealing paddle. This assures a secure uniform seal around all wells. The paddle fits within the rim of a raised-well plate.
Compatible with sealing film 13024-16, 13024-18, and 13024-82. Sealing film sold separately.