Extech 881603 Type K Immersion Probe Stainless Steel
Representative image only.
Extech Mfr # 881603 – Item # 9500154
Choose the type for your unique application
  • Type J/K and RTD available
  • 6 month warranty
PRODUCT OPTIONS
₹11,800.00INR / Each
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel
  • Length (InchShort)2.4375
  • Height (in)1.1093
  • Width (in)15.5
  • Warranty6 month
6 Month Warranty
Key Features
  • Type J/K and RTD available
  • 6 month warranty
More About this Item
Special-application thermocouple probes work with any meter accepting standard thermocouple plug connectors. Choose the Type (J/K) compatible with your meter. Each probe is 39" long.