Extech TP200 K-Type pipe clamp temperature probe with spring-loaded jaw
Representative image only.
Extech Mfr # TP200 – Item # 9500126
Choose the type for your unique application
  • Type J/K and RTD available
  • Applications: FAMEs, pesticides, pharmaceuticals, phenols
  • 1 year warranty
PRODUCT OPTIONS
₹9,900.00INR / Each
₹34,000.00 - ₹43,100.00INR / Each
for an additional
₹34,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Type J/K and RTD available
  • Applications: FAMEs, pesticides, pharmaceuticals, phenols
  • 1 year warranty
More About this Item
Special-application thermocouple probes work with any meter accepting standard thermocouple plug connectors. Choose the Type (J/K) compatible with your meter. Each probe is 39" long.
Type K Pipe Clamp Temperature Probe for hands-free superheat/sub-cooling temperature measurement from
-4°F to 200°F (-20°C to 93C)