Extech TP870 Thermocouple Probe, Beaded Wire, Type K
Representative image only.
Extech Mfr # TP870 – Item # 9041510
Choose the type for your unique application
  • Type J/K and RTD available
  • Type K temperature probe with mini connector
  • Temperature range:-40 to 482-Degrees F (-40 to 250-Degrees C)
  • Type K mini connector
  • Compatible with most meters that have Type K temperature function
  • 6 month warranty
PRODUCT OPTIONS
₹3,900.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Length (InchShort)39
  • Warranty6 month
6 Month Warranty
Key Features
  • Type J/K and RTD available
  • Type K temperature probe with mini connector
  • Temperature range:-40 to 482-Degrees F (-40 to 250-Degrees C)
  • Type K mini connector
  • Compatible with most meters that have Type K temperature function
  • 6 month warranty
More About this Item
Special-application thermocouple probes work with any meter accepting standard thermocouple plug connectors. Choose the Type (J/K) compatible with your meter. Each probe is 39" long.