Fischer Technology 604-142 Thickness Gauge Probe 0 to 45 mils, for models Isoscope FMP10 & Dualscope FMP20
Representative image only.
Fischer Technology Mfr # 604-142 – Item # 5410008
Fischer offers a wide range of probes for a variety of measurement tasks
  • Determine heat damage by means of conductivity measurement
  • Determine ferrite content of steel without destruction
  • Available for sale in the United States, Canada, and Mexico only
  • 6 month warranty
PRODUCT OPTIONS
₹236,000.00INR / Each
₹34,000.00 - ₹43,100.00INR / Each
for an additional
₹34,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Range0 to 45 mil (0 to 1200 µm)
  • Accuracy±0.5 µm up to 50 µm; ±1% from 50 to 800 µm; < 3 % above 800 µm
  • Measuring MethodEddy current
  • ProbeEddy Current
  • DescriptionThickness Gauge Probe 0 to 45 mils, for models Isoscope FMP10 & Dualscope FMP20
  • Warranty6 month
6 Month Warranty
Key Features
  • Determine heat damage by means of conductivity measurement
  • Determine ferrite content of steel without destruction
  • Available for sale in the United States, Canada, and Mexico only
  • 6 month warranty
More About this Item
Choose the appropriate probe for your application.