Fischer Technology 604-175 Probe for Inside measurements, 0 to 40 mils, FGABI1.3-150, for models Deltascope FMP10 & Dualscope
Representative image only.
Fischer Technology Mfr # 604-175 – Item # 5410007
Fischer offers a wide range of probes for a variety of measurement tasks
  • Determine heat damage by means of conductivity measurement
  • Determine ferrite content of steel without destruction
  • 6 month warranty
PRODUCT OPTIONS
₹363,000.00INR / Each
₹34,000.00 - ₹43,100.00INR / Each
for an additional
₹34,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Range 0 to 40 mil (0 to 1000 µm)
  • Accuracy±0.5 µm up to 50 µm; < 1% above 50 µm
  • Measuring MethodMagnetic induction
  • Display240x160 pixels
  • ProbeMagnetic induction
  • DescriptionProbe for Inside measurements, 0 to 40 mils, FGABI1.3-150, for models Deltascope FMP10 & Dualscope
  • Warranty6 month
6 Month Warranty
Key Features
  • Determine heat damage by means of conductivity measurement
  • Determine ferrite content of steel without destruction
  • 6 month warranty
More About this Item
Choose the appropriate probe for your application.