Fischer Technology 604-178 Right angle, single tip, Model FGABW1.3, 0 to 60 mils, for models FMP10
Representative image only.
Fischer Technology Mfr # 604-178 – Item # 5410006
Fischer offers a wide range of probes for a variety of measurement tasks
  • Determine heat damage by means of conductivity measurement
  • Determine ferrite content of steel without destruction
  • Available for sale in the United States, Canada, and Mexico only
  • 6 month warranty
PRODUCT OPTIONS
₹290,000.00INR / Each
₹34,000.00 - ₹43,100.00INR / Each
for an additional
₹34,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Range0 to 80 mil (0 to 2000 µm)
  • Accuracy±1 µm up to 100 µm; ±1% from 100 to 1000 µm; < 3 % above 1000 µm
  • Measuring MethodMagnetic induction
  • DescriptionRight angle, single tip, Model FGABW1.3, 0 to 60 mils, for models FMP10
  • Warranty6 month
6 Month Warranty
Key Features
  • Determine heat damage by means of conductivity measurement
  • Determine ferrite content of steel without destruction
  • Available for sale in the United States, Canada, and Mexico only
  • 6 month warranty
More About this Item
Choose the appropriate probe for your application.