Fisher WLT500 Water Level Indicator 500FT
Representative image only.
Fisher Mfr # WLT500 – Item # 9905016
Contact of the electrode assembly with the water surface is indicated by an LED and the sound of the audio alert beeper
  • Depth indication is provided by numbered metal tags securely crimped to the cable at intervals of five feet
  • 2 year warranty
PRODUCT OPTIONS
₹308,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Depth (ft)500
  • DescriptionWater Level Indicator 500FT
  • Warranty2 year
Two year warranty
Key Features
  • Depth indication is provided by numbered metal tags securely crimped to the cable at intervals of five feet
  • 2 year warranty
More About this Item
The brass electrode assembly is 3/8” in diameter and is easily replaceable in the field. The cable is two-conductor, steel and copper plated, with a tensile strength of approximately 300 lbs and a tough plastic type insulating jacket. It is mounted on a small hand-held spool and stand which can be used conveniently to let out or withdraw the cable and electrode assembly.