Fuego SCS Burner with Infrared Sensor

Representative image only.
Fuego Mfr # 8,200,000 – Item # 3630204
Simple and safe operation for all flame related applications in the laboratory
  • Ideal for use in all industrial, laboratory, and cleanroom settings
  • Modern low-profile body design reduces air flow disturbances and is very easy to clean
  • Robust, all stainless steel construction and fireproof controls
  • 2 year warranty

PRODUCT OPTIONS

₹178,000.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Gas TypeNatural Gas, Propane/Butane
  • Inlet Tube OD (in)0.25
  • Width (in)5 1/2
  • Length (in)4
  • Height (in)1 15/16
  • Power (VAC)120 / 240
  • Warranty2 Years
Two year warrantyUL Listed to Canadian safety standards.Conformite Europeenne

Key Features

  • Ideal for use in all industrial, laboratory, and cleanroom settings
  • Modern low-profile body design reduces air flow disturbances and is very easy to clean
  • Robust, all stainless steel construction and fireproof controls
  • 2 year warranty

More About this Item

With an ergonomic design and extensive standard features, these laboratory gas burners withstand extreme lab conditions while incorporating innovative safety technology. The Safety Control System (SCS) constantly analyzes potential hazards, and, if necessary, initiates safety measures, such as an interruption of the gas supply. An adjustable automatic cut-off system inhibits unintentional ignition. To protect you from burns after prolonged use, the residual heat display will signal that the burner head is still hot. Burners also feature continuous burner head control which identifies burner head clogging and ensures correct assembly of burner head.

Fuego SCS Burner incorporates a touch-free IR sensor and clear, easy-to-read multilanguage LCD. When activated, the unique IR sensor ensures that the burner can only be ignited by clicking the IR sensor twice. Unintentional ignition or ignition due to dropping or falling objects is virtually impossible. The logical menu and display facilitate the fast activation of any function. High-precision burning time regulation and IR sensor reaction distance adjustment are easily possible. Five programs are available, Foot pedal optional.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat