General Tools 80557 LED-Lighted Telescoping Inspection Mirror, Round

Representative image only.
General Tools Mfr # 80557 – Item # 0991405
Two extra-bright LED lights help see into dark and hard-to-reach areas
  • Easily inspect tight areas for leaks, breaks, or lost articles
  • Shaft extends to 33" (840 mm)
  • Large, fully polished, shatter-resistant glass mirrors sit on a 360° swivel
  • Ball joints allow for positioning mirrors at any angle
  • Nonslip cushion grip
  • 1 year warranty

PRODUCT OPTIONS

Call For Price

SPECS & DESCRIPTION

Specifications & Description

  • Mirror TypeRound
  • Mirror Diameter (in)1.875
  • Mirror Diameter (mm)47.63
  • Adjustable Handle Length12" to 33"
  • Warranty1 year
1 Year WarrantyConformite Europeenne

Key Features

  • Easily inspect tight areas for leaks, breaks, or lost articles
  • Shaft extends to 33" (840 mm)
  • Large, fully polished, shatter-resistant glass mirrors sit on a 360° swivel
  • Ball joints allow for positioning mirrors at any angle
  • Nonslip cushion grip
  • 1 year warranty

More About this Item

These lighted inspection mirrors make it easy to inspect tight areas for leaks, breaks, or lost articles. Telescoping shaft extends to 33" (84 cm). Large, fully polished, shatter-resistant glass mirrors sit on a 360° swivel ball joint, allowing mirror to be positioned at any angle. Handle features a nonslip cushion grip.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat