Glas-Col 104A HS112-3 Temperature/Motor Controller, 3 Load Receptacles; 120V

Representative image only.
Glas-Col Mfr # 104A HS112-3 – Item # 3622692
Accurately controls heating mantel speed and temperature
  • Full PID control with universal input
  • Offers three load receptacles for motor drive stirring/heating mantels
  • 1 year warranty

PRODUCT OPTIONS

₹431,000.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Power (VAC)120
  • Power (watts)800
  • DescriptionTemperature/Motor Controller, 3 Load Receptacles; 120V
  • Warranty1 Year
1 Year Warranty

Includes

Power cord.

Key Features

  • Full PID control with universal input
  • Offers three load receptacles for motor drive stirring/heating mantels
  • 1 year warranty

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat