Glass Encased magnetic Stir Bars, 1 1/2"l X 3/8"d

Representative image only.
Item # 0476953
  • Borosilicate glass-encased magnetic stir bars are completely non-reactive, leak proof, and nonabsorbent
  • Handles temperatures up to 525°F (274°C)

PRODUCT OPTIONS

₹6,100.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Length (in)1 1/2
  • Length (cm)3.81
  • Diameter (in)0.375
  • Diameter (cm)0.9525

Key Features

  • Borosilicate glass-encased magnetic stir bars are completely non-reactive, leak proof, and nonabsorbent
  • Handles temperatures up to 525°F (274°C)

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat