Humboldt H-4200 Stainless Steel Soil Pocket Penetrometer; 0 to 4.5 tons per sq. ft

Representative image only.
Humboldt Mfr # H-4200 – Item # 9903910
Lightweight, spring-operated penetrometer quickly measures the compressive soil strength
  • Easy to use with easy-to-read results
  • THE FOOT ADAPTER IS NOT INCLUDED BUT IS AVAILABLE SEPARATELY
  • 1 year warranty

₹26,900.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • MaterialStainless Steel
  • Core Diameter (in)21/32
  • Core Diameter (cm)160
  • Length (in)7 3/4
  • Length (cm)20
  • Warranty1 year
1 Year Warranty

Key Features

  • Easy to use with easy-to-read results
  • THE FOOT ADAPTER IS NOT INCLUDED BUT IS AVAILABLE SEPARATELY
  • 1 year warranty

More About this Item

Penetrometer is easy to use: push the scale into the penetrometer body until the 0.0 mark is showing. Inset stainless steel piston into soil until indicating mark. The easy-to-read scale indicates strength from 0 to 4.5 tons per sq ft. The reading is maintained until manually reset.

Penetrometer is constructed from stainless steel and measures 7-3/4"L x 2/3" dia (200 x 16 mm).

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat