Idex 316 Stainless Steel Nut, Waters Compatible, 1/16" OD Tubing, 10-32 Coned; 10/PK
Representative image only.
Idex Mfr # U-410X – Item # 0201474
Stainless steel fittings withstand pressures up to 20,000 psi (1380 bar)!
  • Constructed of durable stainless steel
  • Waters Compatible
  • 10-32 coned configuration
  • 10/PK
  • 1 month warranty
  • 1 month warranty
PRODUCT OPTIONS
₹13,600.00INR / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialStainless steel
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 1) (in)10-32 coned
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Compression Size / Tube Od (Port 1) (mm)1.588
  • Max Operating Pressure (PSI)20000
1 Month Warranty
Key Features
  • Constructed of durable stainless steel
  • Waters Compatible
  • 10-32 coned configuration
  • 10/PK
  • 1 month warranty
  • 1 month warranty
More About this Item
These 316 stainless steel nuts (and ferrules) are rated to 20,000 psi (1380 bar) when tightened using a wrench. Recommended torque for tightening these fittings is 20 in-lbs. (2.25 N-m).
Metal fittings should not be used with plastic ports because they can damage the ports.