Idex Lock Nut for 1/4-28 Flangeless Fittings, White Acetal; 1/EA
Representative image only.
Idex Mfr # P-312 – Item # 0201996
Hold flangeless fittings securely in place
  • Optimize performance of your flangeless fittings
  • 1/EA
  • 1 year warranty
PRODUCT OPTIONS
₹890.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialDelrin® acetal
  • Thread size UNF(M) (port 1) (in)1/4-28 flat bottom
  • Warranty1 year
1 Year Warranty
Key Features
  • Optimize performance of your flangeless fittings
  • 1/EA
  • 1 year warranty
More About this Item
These lock nuts are ideal for applications where vibrations can loosen fittings