Idex M-520 Inline Microfilter Assembly, SS, 0.5 µm PEEK Frit, MicroTight® Tubing Sleeves; 1/EA

Representative image only.
Idex Mfr # M-520 – Item # 4271131
Protect your columns from particulate contamination
  • Withstands up to 5000 psi (345 bar)
  • Use with MicroTight® tubing sleeves
  • PEEK frit
  • 1 month warranty

PRODUCT OPTIONS

₹34,600.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Membrane MaterialPEEK
  • Housing MaterialStainless Steel
  • Pore Size (µm)0.5
  • ConnectionsMicroTight® tubing sleeves
  • Max Pressure (PSI)4000
1 Month Warranty

Includes

Five end fittings 42711-23 and two micro fingertight PEEK nuts 02013-42

Key Features

  • Withstands up to 5000 psi (345 bar)
  • Use with MicroTight® tubing sleeves
  • PEEK frit
  • 1 month warranty

More About this Item

Specifically engineered inline solvent MicroFilters prevent particulate contamination ensuring reliable chromatographic results even at nanoliter flow rates. Filters protect columns from particles often originating in the mobile phase or sample, or from pump seal and sample injection valve wear. Connect directly using 1/32" OD tubing or choose MicroTight® tubing sleeve models for use with capillary tubing.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat