IKA H 135.103 Block 9 x 16 mL, 3.1" x 3.1".
Representative image only.
IKA Mfr # 0025000628 – Item # 0467203
Perform multiple synthesis operations with these expansion blocks, flasks and inlays.
  • The aluminum blocks guarantee optimal heat transfer throughout the process without interference to the magnetic field.
  • Exchangeable flask inserts provide high-precision thermal conduction directly into the flask.
  • 2 year warranty
PRODUCT OPTIONS
₹8,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithSynthesis Package 1
  • Warranty2 year
Two year warranty
Key Features
  • The aluminum blocks guarantee optimal heat transfer throughout the process without interference to the magnetic field.
  • Exchangeable flask inserts provide high-precision thermal conduction directly into the flask.