IKA SAS-1 Spiral Adapter
Representative image only.
IKA Mfr # 25000377 – Item # 8305647
Extensive accessory options for IKA® ROTAVISC viscometers
  • Optional spindles
  • Adapters
  • Software
  • Measures the viscosity of very sticky samples
  • Possible to analyse the fluid behavior of different shear rates
  • 2 year warranty
PRODUCT OPTIONS
₹397,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSAS-1 Spiral Adapter
Two year warranty
Key Features
  • Optional spindles
  • Adapters
  • Software
  • Measures the viscosity of very sticky samples
  • Possible to analyse the fluid behavior of different shear rates
  • 2 year warranty
More About this Item
Different accessories facilitate the viscosity reading of different types of fluids. Choose the accessories that best fit your application.