IKA T-Spindle Set
Representative image only.
IKA Mfr # 25007765 – Item # 8305553
Extensive accessory options for IKA® ROTAVISC viscometers
  • Optional spindles
  • Adapters
  • Software
  • Particularly suitable for the measurement of non-flowing samples, such as creams, pastes or gelatin
  • 2 year warranty
PRODUCT OPTIONS
₹236,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionT-Spindle Set
Two year warranty
Includes
6 T-spindles, nut, coupling, and carrying case.
Key Features
  • Optional spindles
  • Adapters
  • Software
  • Particularly suitable for the measurement of non-flowing samples, such as creams, pastes or gelatin
  • 2 year warranty
More About this Item
Different accessories facilitate the viscosity reading of different types of fluids. Choose the accessories that best fit your application.