IKA VOL-C-RTD-2.1 Optional Chamber
Representative image only.
IKA Mfr # 25007064 – Item # 8305669
Extensive accessory options for IKA® ROTAVISC viscometers
  • Optional spindles
  • Adapters
  • Software
  • Especially suitable for expensive and valuable samples
  • A temperature sensor is already integrated in the chamber and thus allows for temperature measurement during viscosity determination
  • 2 year warranty
PRODUCT OPTIONS
Loading
₹196,000.00INR / Each
Loading
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionVOL-C-RTD-2.1 Optional Chamber
Two year warranty
Key Features
  • Optional spindles
  • Adapters
  • Software
  • Especially suitable for expensive and valuable samples
  • A temperature sensor is already integrated in the chamber and thus allows for temperature measurement during viscosity determination
  • 2 year warranty
More About this Item
Different accessories facilitate the viscosity reading of different types of fluids. Choose the accessories that best fit your application.