InnoCal A2LA ISO17025 Accredited Calibration; Dewpoint ≥ -40°C, 5 Points

Representative image only.
InnoCal Item # 1750025
Supports calibrations for all types of thermohygrometers
  • Calibrates hygrometers with 5 to 95% relative humidity and dew point as low as -70°C
  • Services all major brands including Cole-Parmer, Digi-Sense, Extech, Fluke, Oakton, and Vaisala
  • Receive your product calibrated and ready to use—eliminates delays by third-party or internal calibration labs
  • NIST-traceable calibration confirms your purchase meets the manufacturer’s published specification

PRODUCT OPTIONS

₹50,300.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Calibration TypeA2LA ISO17025 Accredited Humidity
  • System Calibration (Instrument and Probe)Yes
  • Number Of Calibration Test Points3 humidity, 1 temperature, 5 dewpoint readings
  • Calibration Test PointsHumidity points across the range and dewpoint down to -40°C (5 points)
  • Calibration Test Range5 to 95% RH, dewpoint to -40°C
  • DescriptionA2LA ISO17025 Accredited Calibration; Dewpoint ≥ -40°C, 5 Points

Key Features

  • Calibrates hygrometers with 5 to 95% relative humidity and dew point as low as -70°C
  • Services all major brands including Cole-Parmer, Digi-Sense, Extech, Fluke, Oakton, and Vaisala
  • Receive your product calibrated and ready to use—eliminates delays by third-party or internal calibration labs
  • NIST-traceable calibration confirms your purchase meets the manufacturer’s published specification

More About this Item

All calibration certificates contain as-found and as-left data, estimated measurement uncertainties (EMU), and test uncertainty ratios (TUR) at no additional costs!
EPA, USDA, ISO and GLP/GMPs standards require regular NIST-traceable calibration of all test equipment being used in critical applications. Measurements reported on our reports are traceable to SI units via national standards maintained by NIST and are performed in compliance with ANSI Z540, ISO17025, and MIL-STD-45662A. The results stated in our reports relate only to the items tested or calibrated.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat