InnoCal NIST-Traceable Calibration; Tachometer
Representative image only.
InnoCal Item # 1705000
  • Calibration service for contact tachometers (mechanical), non-contact tachometers (optical), and combination tachometers featuring both contact and non-contact options
PRODUCT OPTIONS
₹34,000.00INR / Each
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • System Calibration (Instrument and Probe)Yes
  • Calibration Test PointsThroughout range of instrument
  • Calibration Test RangeMechanical: 60 to 24,000 rpm; Optical: 0 to 200,000 rpm
Key Features
  • Calibration service for contact tachometers (mechanical), non-contact tachometers (optical), and combination tachometers featuring both contact and non-contact options
More About this Item
Calibration reports issued by InnoCal may not be reproduced, except in full, without written permission of InnoCal. The results stated in our reports relate only to the items tested or calibrated. Measurements reported on our reports are traceable to SI units via national standards maintained by NIST and are performed in compliance with MIL-STD-45662A, ANSI/NCSL Z540-1-1994, 10CFR50, Appendix B, ISO9002-94, and ISO/IEC 17025:2005
InnoCal's Metrology Laboratory is accredited to ISO/IEC 17025 by the American Association for Laboratory Accreditation (A2LA); certificate number 1746.01. This accreditation is your assurance that one of the foremost outside auditing agencies approves of our internal laboratory policies and procedures.