Jenway 025 196 pH Electrode Fill Solution, 4M KCl, AgCl Saturated, 150 mL

Representative image only.
Jenway Item # 9995961
All your pH measurement needs in one place
  • Jenway offers a range of pH buffers either as single use sachets or longer lasting 500 mL bottles

PRODUCT OPTIONS

₹11,700.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Accessory TypepH Electrode Fill Solution
  • Range4M KCl, AgCl Saturated; 150 mL
  • DescriptionpH Electrode Fill Solution, 4M KCl, AgCl Saturated; 150 mL

Key Features

  • Jenway offers a range of pH buffers either as single use sachets or longer lasting 500 mL bottles

More About this Item

To care for your Jenway pH electrodes there is also a range of storage solutions and fill solutions tailored to your specific probe.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat