Jenway 027 227 Waterproof, General Purpose, Epoxy-Bodied pH Electrode with ATC
Representative image only.
Jenway Item # 5953070
A variety of pH electrodes for your Jewnay meters
  • Combination electrodes
  • Application specific electrodes
  • Waterproof electrodes
  • Temperature and reference electrodes
  • 1 year warranty
PRODUCT OPTIONS
₹18,700.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsStandard measurements of general solutions
  • Electrode TypeCombination
  • Electrode Body MaterialEpoxy
  • Reference JunctionCeramic PTFE
  • Reference ElectrolyteAgCl
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Temperature CompensationYes
  • Connector3150/3205 Jenway meter waterproof connector
  • Diameter (mm)12
  • Length (mm)120
  • Cable Length (m)1
  • DescriptionWaterproof, General Purpose, Epoxy-Bodied pH Electrode with ATC
1 Year Warranty
Key Features
  • Combination electrodes
  • Application specific electrodes
  • Waterproof electrodes
  • Temperature and reference electrodes
  • 1 year warranty
More About this Item
Choose the right electrode for your application.