Jenway 924 017 Double Junction, AgCl, Glass Bodied Reference Electrode
Representative image only.
Jenway Item # 7900075
A variety of pH electrodes for your Jewnay meters
  • Combination electrodes
  • Application specific electrodes
  • Waterproof electrodes
  • Temperature and reference electrodes
PRODUCT OPTIONS
₹14,200.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsFor use with any mono electrode
  • Electrode TypeReference
  • Electrode Body MaterialGlass
  • Reference JunctionCeramic frit
  • Reference ElectrolyteAgCl
  • Min Temperature (° C)0
  • Max Temperature (° C)50
  • Connector2 mm plug
  • Diameter (mm)12
  • Length (mm)120
  • Cable Length (m)1
  • DescriptionDouble Junction, AgCl, Glass Bodied Reference Electrode
Key Features
  • Combination electrodes
  • Application specific electrodes
  • Waterproof electrodes
  • Temperature and reference electrodes
More About this Item
Choose the right electrode for your application.