Jenway 924 068 Environmental pH Electrode with S7 Connector
Representative image only.
Jenway Item # 7900080
A variety of pH electrodes for your Jewnay meters
  • Combination electrodes
  • Application specific electrodes
  • Waterproof electrodes
  • Temperature and reference electrodes
PRODUCT OPTIONS
₹22,300.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsLow temperatures or solutions of low ionic strength
  • Electrode TypeCombination
  • Electrode Body MaterialGlass
  • Reference JunctionAnnular ceramic
  • Reference ElectrolyteHgCl
  • Min pH (pH)0
  • Max pH (pH)11
  • Min Temperature (° C)0
  • Max Temperature (° C)50
  • ConnectorS7
  • Diameter (mm)12
  • Length (mm)120
  • Cable Length (m)1
  • DescriptionEnvironmental pH Electrode with S7 Connector
Key Features
  • Combination electrodes
  • Application specific electrodes
  • Waterproof electrodes
  • Temperature and reference electrodes
More About this Item
Choose the right electrode for your application.