Kanomax Airflow Measurement Probe for Velocity and Temperature; Omnidirectional, Sphere
Representative image only.
Kanomax Mfr # 6543-2G – Item # 1000728
Research-grade accuracy for airflow measurement—ideal for laboratory, IAQ, and critical environment monitoring
  • Smart probe technology lets you change probes without recalibrating the main unit—order meter separately to create your system
  • 1 year warranty
PRODUCT OPTIONS
Loading
₹251,000.00INR / Each
Loading
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)-4
  • Max Temperature (° F)158
  • Min Temperature (° C)-20
  • Max Temperature (° C)70
  • DescriptionAirflow Measurement Probe for Velocity and Temperature; Omnidirectional, Sphere
  • Warranty1 year
1 Year Warranty
More About this Item
Unidirectional probes are economical and great for applications where the direction of the airflow is known. Omnidirectional probes can sense airflow from all directions—perfect for turbulent airflow applications.