Kanomax Airflow Measurement Probe for Velocity; Omnidirectional, Mini L Shape SKU 1000729
Representative image only.
Kanomax Mfr # 6551-2G – Item # 1000729
Research-grade accuracy for airflow measurement—ideal for laboratory, IAQ, and critical environment monitoring
  • Smart probe technology lets you change probes without recalibrating the main unit—order meter separately to create your system
  • 1 year warranty
PRODUCT OPTIONS
₹361,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAirflow Measurement Probe for Velocity; Omnidirectional, Mini L Shape
  • Warranty1 year
1 Year Warranty
Key Features
  • Smart probe technology lets you change probes without recalibrating the main unit—order meter separately to create your system
  • 1 year warranty
More About this Item
Unidirectional probes are economical and great for applications where the direction of the airflow is known. Omnidirectional probes can sense airflow from all directions—perfect for turbulent airflow applications.