Kimberly-Clark Kimtech Kimwipes Ex-l Wipes and Dispenser Kit, 4.5" X 8.5"; 280/Bx; 6 Boxes/Cs
Representative image only.
Kimberly-Clark Mfr # MAKE-UP – Item # 2500039
Bundled for convenience at a bargain price!
  • Kit includes mountable wipe dispenser and six boxes of wipes
  • Low-lint critical task wipes are absorbent, soft, and nonabrasive
  • Dispenser is simple to mount—peel adhesive tape and press onto surface
  • Wipe dispenser is constructed of durable white PVC plastic
₹10,700.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Width (in)4 1/2
  • Length (in)8 1/2
  • DescriptionWipes and Dispenser Kit, 4.5" X 8.5"; 280/Bx; 6 Boxes/Cs
Includes
Key Features
  • Kit includes mountable wipe dispenser and six boxes of wipes
  • Low-lint critical task wipes are absorbent, soft, and nonabrasive
  • Dispenser is simple to mount—peel adhesive tape and press onto surface
  • Wipe dispenser is constructed of durable white PVC plastic
More About this Item
Wipes won’t scratch delicate surfaces and they wipe surfaces clean without leaving residue. Single-ply Kimtech Science Kimwipes® Ex-l delicate task wipers are made of a finely textured tissue, and come in a Pop-Up® box with antistatic Lintguard® polyshield — minimizing lint, static buildup, and contamination.
The wipe dispenser holds a single box of wipes and is ready to mount near instruments, workstations, hoods, and benches. Free up valuable drawer and bench space — peel off the adhesive tape backing and press onto surface.